missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-O-methyl-L-threonine, 95%, Thermo Scientific™
£27.50 - £251.00
Chemical Identifiers
CAS | 48068-25-3 |
---|---|
Molecular Formula | C10H19NO5 |
Molecular Weight (g/mol) | 233.26 |
MDL Number | MFCD00076990 |
InChI Key | VWSUOKFUIPMDDX-RQJHMYQMSA-N |
Synonym | boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid |
PubChem CID | 15460346 |
IUPAC Name | (2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
SMILES | CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
48068-25-3 | |
233.26 | |
VWSUOKFUIPMDDX-RQJHMYQMSA-N | |
15460346 | |
CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O |
C10H19NO5 | |
MFCD00076990 | |
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid | |
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid |
Specifications
48068-25-3 | |
MFCD00076990 | |
VWSUOKFUIPMDDX-RQJHMYQMSA-N | |
(2S,3R)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid | |
15460346 | |
95% | |
N-Boc-O-methyl-L-threonine |
C10H19NO5 | |
boc-o-methyl-l-threonine, boc-thr me-oh, 2s,3r-2-tert-butoxycarbonylamino-3-methoxybutanoic acid, 2s,3r-2-tert-butoxycarbonyl amino-3-methoxybutanoic acid, n-boc-o-methyl-l-threonine, o-methyl-l-threonine, n-boc protected, n-tert-butoxycarbonyl-o-methyl-l-threonine, n-tert-butyloxycarbonyl-o-methyl-l-threonine, 2s,3r-2-tert-butoxy carbonyl amino-3-methoxybutanoic acid | |
CO[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
233.26 | |
233.26 | |
Solid |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold