missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-(4-methoxybenzyl)-L-cysteine, 98%, Thermo Scientific™

Product Code. 15470699
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18942-46-6
Molecular Formula C16H23NO5S
Molecular Weight (g/mol) 341.42
MDL Number MFCD00038254
InChI Key VRTXRNJMNFVTOM-UHFFFAOYNA-N
Synonym boc-cys pmeobzl-oh, boc-s-4-methoxybenzyl-l-cysteine, boc-cys 4-meobzl-oh, boc-cys mob-oh, r-2-tert-butoxycarbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-tert-butoxycarbonyl-s-4-methoxybenzyl-l-cysteine, 2r-2-tert-butoxycarbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, n-tert-butoxycarbonyl-s-p-methoxybenzyl-l-cysteine, l-cysteine, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl, n-1,1-dimethylethoxy carbonyl-s-4-methoxyphenyl methyl-l-cysteine
PubChem CID 87863
IUPAC Name (2R)-3-[(4-methoxyphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES COC1=CC=C(CSCC(NC(=O)OC(C)(C)C)C(O)=O)C=C1

Specifications

Formula Weight 341.42
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material N-Boc-S-(4-methoxybenzyl)-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'