missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-thiourea, 97%, Thermo Scientific Chemicals

Product Code. 15410379
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-thiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 268551-65-1
Molecular Formula C6H12N2O2S
Molecular Weight (g/mol) 176.23
MDL Number MFCD04039987
InChI Key JKXQTODLIZBUDE-UHFFFAOYSA-N
Synonym n-boc-thiourea, n-tert-butoxycarbonylthiourea, n-boc thiourea, mono-boc-thiourea, acmc-20ak5u, n-tert-butoxycarbonyl-thiourea, n-tert.butoxycarbonyl thiourea, 1-tert-butoxycarbonyl thiourea, tert-butyl carbamothioylcarbamate
PubChem CID 4095660
IUPAC Name tert-butyl N-carbamothioylcarbamate
SMILES CC(C)(C)OC(=O)NC(N)=S

Specifications

Melting Point 142°C (decomposition)
Color White
Formula Weight 176
Percent Purity 97%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Boc-thiourea

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'