missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-trans-4-hydroxy-L-proline benzyl ester, 95%, Thermo Scientific Chemicals

Product Code. 15447496
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89813-47-8
Molecular Formula C17H23NO5
Molecular Weight (g/mol) 321.373
MDL Number MFCD00076979
InChI Key BEIPCYKSYYZEJH-KGLIPLIRSA-N
Synonym boc-hyp-obzl, boc-o-benzyl-l-hydroxyproline, 2s,4r-2-benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate, 2-benzyl 1-tert-butyl 2s,4r-4-hydroxypyrrolidine-1,2-dicarboxylate, bochypobn, boc-hyp-obn, 2s,4r-2-benzyl 1-t-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
PubChem CID 11902907
IUPAC Name 2-O-benzyl 1-O-tert-butyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OCC2=CC=CC=C2)O

Specifications

Color Brown
Formula Weight 321.37
Percent Purity 95%
Quantity 250 mg
Physical Form Liquid
Chemical Name or Material N-Boc-trans-4-hydroxy-L-proline benzyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'