missing translation for 'onlineSavingsMsg'
Learn More

N-Bromosuccinimide, 99%

Product Code. 10655332 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
10g
1kg
500g
5kg
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10655332

Brand: Thermo Scientific Acros 107451000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 128-08-5 | C4H4BrNO2 | 177.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent, C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-08-5
MDL Number MFCD00005510
InChI Key PCLIMKBDDGJMGD-UHFFFAOYSA-N
Synonym n-bromosuccinimide, succinbromimide, succinbromide, succinibromimide, n-bromosuccimide, 2,5-pyrrolidinedione, 1-bromo, 1-bromo-2,5-pyrrolidinedione, bromosuccinimide, nbs, n-bromo succinimide
PubChem CID 67184
ChEBI CHEBI:53174
IUPAC Name 1-bromopyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White to Yellow
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H4BrNO2
Quantity 100 g
Beilstein 21, 380
Fieser 01,78; 02,40; 03,34; 04,49; 05,65; 06,74; 07,37; 08,54; 09,70; 10,57; 11,79; 12,79; 13,41; 14,57; 15,50; 16,49
Merck Index 15, 1448
Molecular Weight (g/mol) 177.985
Formula Weight 177.99
Percent Purity 98.5 to 101.5% (on dry substance)
Physical Form Crystalline Powder
Chemical Name or Material N-Bromosuccinimide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-Bromosuccinimide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.