missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals

Product Code. 11477936
250 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.

Solubility
Soluble in DMSO, ethyl acetate, methanol.

Notes
Store in refrigerator. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-26-8
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00016478
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
Synonym butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben, butyl chemosept, butyl parasept, butyl tegosept, butyl butex, tegosept butyl
PubChem CID 7184
IUPAC Name butyl 4-hydroxybenzoate
SMILES CCCCOC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 67°C to 71°C
Quantity 250 g
Beilstein 1103741
Merck Index 14,1584
Solubility Information Soluble in DMSO,ethyl acetate,methanol.
Formula Weight 194.23
Percent Purity ≥99%
Chemical Name or Material n-Butyl 4-hydroxybenzoate
compliance-icons
Product Identifier
  • n-Butyl 4-hydroxybenzoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : n-Butyl 4-hydroxybenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Butyl 4-hydroxybenzoate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.