Learn More
Chemical Identifiers
CAS | 123-86-4 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00009445 |
InChI Key | DKPFZGUDAPQIHT-UHFFFAOYSA-N |
Synonym | n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle |
PubChem CID | 31272 |
ChEBI | CHEBI:31328 |
IUPAC Name | butyl acetate |
SMILES | CCCCOC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404017
|
Thermo Scientific Alfa Aesar
A19412.AP |
500 mL |
£19.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414017
|
Thermo Scientific Alfa Aesar
A19412.0F |
2500 mL |
£48.74
2500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In pharmaceutical and petroleum industries, it is used for extraction and as a processing solvent. It is an ingredient of cleaners, essences and fragrances. It is used as an analytical reagent to verify thalium, stannum and tungsten as well as in the determination of molybdenum and rhenium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Butyl acetate is widely used as a solvent for cellulose nitrate, resins, polymers, oils and fats. It is used in paints, coatings, printing inks, aerosol sprays and cosmetics. It is used as a synthetic fruit flavoring agent in foods like candy, ice cream, cheeses and baked food items. In pharmaceutical and petroleum industries, it is used for extraction and as a processing solvent. It is an ingredient of cleaners, essences and fragrances. It is used as an analytical reagent to verify thalium, stannum and tungsten as well as in the determination of molybdenum and rhenium.
Solubility
Miscible with ethanol, ethyl ether, alcohol, acetone, chloroform and most hydrocarbons. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
Chemical Identifiers
123-86-4 | |
116.16 | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
31272 | |
butyl acetate |
C6H12O2 | |
MFCD00009445 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
CHEBI:31328 | |
CCCCOC(C)=O |
Specifications
123-86-4 | |
0.881 | |
124°C to 126°C | |
Fruit-like | |
CH3CO2(CH2)3CH3 | |
MFCD00009445 | |
1741921 | |
n-butyl acetate, acetic acid, butyl ester, 1-butyl acetate, butyl ethanoate, acetic acid butyl ester, n-butyl ethanoate, n-butylacetate, butylacetat, acetic acid n-butyl ester, acetate de butyle | |
DKPFZGUDAPQIHT-UHFFFAOYSA-N | |
butyl acetate | |
31272 | |
116.16 | |
n-Butyl acetate |
-78°C | |
5 | |
25°C (77°F) | |
C6H12O2 | |
1.3945 | |
UN1123 | |
14,1535 | |
Miscible with ethanol,ethyl ether,alcohol,acetone,chloroform and most hydrocarbons. Slightly miscible with water. | |
CCCCOC(C)=O | |
116.16 | |
CHEBI:31328 | |
≥99% |
Safety and Handling
GHS H Statement
H226-H336
Flammable liquid and vapour.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL ACETATES
missing translation for 'einecsNumber' : 204-658-1
missing translation for 'rtecsNumber' : AF7350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- n-Butyl acetate
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : n-Butyl acetate
RUO – Research Use Only