missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals
38.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Color | Colorless to Yellow |
Density | 0.7900g/mL |
Infrared Spectrum | Authentic |
Assay Percent Range | 98+% |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)3NC |
Refractive Index | 1.3950 to 1.397 |
Quantity | 1 g |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS06_Toxic.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
Product Identifier
- n-Butyl isocyanide
- Danger
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H311-Toxic in contact with skin.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'