Learn More
n-Butyl isocyanide, 98+%, Thermo Scientific Chemicals
£53.40 - £168.00
Chemical Identifiers
CAS | 2769-64-4 |
---|---|
Molecular Formula | C5H9N |
Molecular Weight (g/mol) | 83.13 |
MDL Number | MFCD00000008 |
InChI Key | FSBLVBBRXSCOKU-UHFFFAOYSA-N |
Synonym | n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyano |
PubChem CID | 76008 |
IUPAC Name | 1-isocyanobutane |
SMILES | CCCC[N+]#[C-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10317240
|
Thermo Scientific Acros
158380010 |
1 g | Glass bottle |
£53.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607132
|
Thermo Scientific Acros
158380050 |
5 g | Glass bottle |
£168.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2769-64-4 | |
83.13 | |
FSBLVBBRXSCOKU-UHFFFAOYSA-N | |
76008 | |
CCCC[N+]#[C-] |
C5H9N | |
MFCD00000008 | |
n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyano | |
1-isocyanobutane |
Specifications
2769-64-4 | |
100.0 | |
0.7900g/mL | |
98+% | |
CH3(CH2)3NC | |
MFCD00000008 | |
0.79 | |
FSBLVBBRXSCOKU-UHFFFAOYSA-N | |
1-isocyanobutane | |
76008 | |
98+% | |
n-Butyl isocyanide, 98+% |
98.0 | |
Colorless to Yellow | |
Authentic | |
C5H9N | |
1.3950 to 1.397 | |
11,99; 14,63 | |
n-butyl isocyanide, butyl isocyanide, 1-butylisocyanide, n-butylisocyanide, bu-tylisonitrile, n-butylisonitrile, n-butyl isonitrile, butane,1-isocyano | |
CCCC[N+]#[C-] | |
83.13 | |
83.13 | |
Liquid |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-458-7
- n-Butyl isocyanide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H331-Toxic if inhaled.
- H311-Toxic in contact with skin.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only