missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl phenyl ether, 99%

Product Code. 11381677
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11381677

Brand: Thermo Scientific Alfa Aesar L04703.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1126-79-0 | C10H14O | 150.221 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1126-79-0
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00009438
InChI Key YFNONBGXNFCTMM-UHFFFAOYSA-N
Synonym butyl phenyl ether, n-butyl phenyl ether, benzene, butoxy, ether, butyl phenyl, phenyl butyl ether, butoxyphenyl, n-butoxybenzene, butyloxy benzene, n-butylphenylether, butoxy-benzene
PubChem CID 14311
IUPAC Name butoxybenzene
SMILES CCCCOC1=CC=CC=C1

Specifications

Melting Point -19°C
Density 0.938
Boiling Point 210°C to 211°C
Flash Point 82°C (179°F)
Refractive Index 1.497
Quantity 25 g
Beilstein 1635559
Formula Weight 150.22
Percent Purity 99%
Chemical Name or Material n-Butyl phenyl ether

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.