missing translation for 'onlineSavingsMsg'
Learn More

n-Butylamine, 99+%, Thermo Scientific Chemicals

Product Code. 11915171
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-73-9
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.13
MDL Number MFCD00011690
InChI Key HQABUPZFAYXKJW-UHFFFAOYSA-N
Synonym butylamine, n-butylamine, 1-butanamine, 1-aminobutane, 1-butylamine, monobutylamine, n-butylamin, mono-n-butylamine, norvalamine, 1-aminobutan
PubChem CID 8007
ChEBI CHEBI:43799
IUPAC Name butan-1-amine
SMILES CCCCN

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -49.0°C
Density 0.7400g/mL
Boiling Point 78.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Metal drum
Linear Formula CH3(CH2)3NH2
Refractive Index 1.4000 to 1.4020
Quantity 10 L
Beilstein 04,156
Fieser 04,53
Merck Index 15,1545
Specific Gravity 0.74
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol and ether
Viscosity 0.5 mPa.s (20°C)
Formula Weight 73.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material n-Butylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Butylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'