Learn More
n-Butylbenzene, 99%, Thermo Scientific Chemicals
£29.40 - £322.00
Chemical Identifiers
CAS | 104-51-8 |
---|---|
Molecular Formula | C10H14 |
Molecular Weight (g/mol) | 134.22 |
MDL Number | MFCD00009463 |
InChI Key | OCKPCBLVNKHBMX-UHFFFAOYSA-N |
Synonym | n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b |
PubChem CID | 7705 |
ChEBI | CHEBI:44194 |
IUPAC Name | butylbenzene |
SMILES | CCCCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404293
|
Thermo Scientific Alfa Aesar
A10910.18 |
50 g |
£29.40
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414293
|
Thermo Scientific Alfa Aesar
A10910.30 |
250 g |
£93.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494283
|
Thermo Scientific Alfa Aesar
A10910.0B |
1000 g |
£322.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
n-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsn-Butylbenzene is an organic solvent that has been used to induce cell death in vitro and for bioconversion.
Solubility
Sparingly soluble in water (0.012g/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Storage temperature 2 - 8°C. Incompatible with strong oxidizing agents.
Chemical Identifiers
104-51-8 | |
134.22 | |
OCKPCBLVNKHBMX-UHFFFAOYSA-N | |
7705 | |
butylbenzene |
C10H14 | |
MFCD00009463 | |
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b | |
CHEBI:44194 | |
CCCCC1=CC=CC=C1 |
Specifications
104-51-8 | |
0.86 | |
59°C (138°F) | |
C10H14 | |
MFCD00009463 | |
1903395 | |
n-butylbenzene, 1-phenylbutane, benzene, butyl, 1-butylbenzene, butyl-benzene, butane, phenyl, unii-s8xz2901rz, butylbenzene, analytical standard, n-butyl benzene, n4b | |
OCKPCBLVNKHBMX-UHFFFAOYSA-N | |
butylbenzene | |
7705 | |
134.22 | |
n-Butylbenzene |
-88°C | |
181°C to 183°C | |
Aromatic | |
1.489 | |
UN2709 | |
14,1549 | |
Sparingly soluble in water (0.012g/L). | |
CCCCC1=CC=CC=C1 | |
134.22 | |
CHEBI:44194 | |
99% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
missing translation for 'einecsNumber' : 203-209-7
missing translation for 'rtecsNumber' : CY9070000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only