Learn More
n-Butylboronic acid, 98%, Thermo Scientific Chemicals
£18.39 - £137.00
Chemical Identifiers
CAS | 4426-47-5 |
---|---|
Molecular Formula | C4H11BO2 |
Molecular Weight (g/mol) | 101.94 |
MDL Number | MFCD00002106 |
InChI Key | QPKFVRWIISEVCW-UHFFFAOYSA-N |
Synonym | n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane |
PubChem CID | 20479 |
IUPAC Name | butylboronic acid |
SMILES | CCCCB(O)O |
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Packaging | Prezzo | Quantità e disponibilità | ||||
10337471
|
Thermo Scientific Acros
172380010 |
1 g | Glass bottle |
£18.39
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10786822
|
Thermo Scientific Acros
172380050 |
5 g | Glass bottle |
£38.59
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10056043
|
Thermo Scientific
172380250 |
25 g | Glass bottle |
£137.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagentIdentificatori chimici
4426-47-5 | |
101.94 | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
20479 | |
CCCCB(O)O |
C4H11BO2 | |
MFCD00002106 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
butylboronic acid |
Specifica
4426-47-5 | |
100.0 | |
Colorless to Yellow | |
98% | |
CH3(CH2)3B(OH)2 | |
n-butylboronic acid, 1-butylboronic acid, 1-butaneboronic acid, n-butaneboronic acid, boronic acid, butyl, 1-butane boronic acid, butylboric acid, butyl boronic acid, chembl31962, 1-butyldihydroxyborane | |
QPKFVRWIISEVCW-UHFFFAOYSA-N | |
butylboronic acid | |
20479 | |
98% | |
n-Butylboronic acid |
97.5 | |
94.0°C to 96.0°C | |
Authentic | |
C4H11BO2 | |
MFCD00002106 | |
Solubility in water:+/- 2.5% in water | |
CCCCB(O)O | |
101.94 | |
101.94 | |
Liquid |
Sicurezza e movimentazione
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 224-607-7
- n-Butylboronic acid
Avvertenza
- Warning
Categoria di pericolo
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Indicazioni di pericolo
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Consigli di prudenza
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only