missing translation for 'onlineSavingsMsg'
Learn More

n-Butyronitrile, 99%, Thermo Scientific Chemicals

Product Code. 10577743
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-74-0
MDL Number MFCD00001968
InChI Key KVNRLNFWIYMESJ-UHFFFAOYSA-N
Synonym butyronitrile, n-butyronitrile, propyl cyanide, 1-cyanopropane, n-propyl cyanide, butyrylonitrile, butyric acid nitrile, n-butanenitrile, propylkyanid, butane nitrile
PubChem CID 8008
ChEBI CHEBI:51937
IUPAC Name butanenitrile
SMILES CCCC#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -112.0°C
Density 0.7900g/mL
Boiling Point 115.0°C to 117.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7N
Linear Formula CH3CH2CH2CN
Refractive Index 1.3830 to 1.3850
Quantity 100 mL
Beilstein 02, II, 252
Merck Index 15, 1601
Specific Gravity 0.79
Solubility Information Solubility in water: 33g/L (25°C). Other solubilities: miscible with alcohol,ether and dimethylformamide
Molecular Weight (g/mol) 69.11
Viscosity 0.57 mPa.s (20°C)
Formula Weight 69.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Butyronitrile
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.