missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Carbamoyl-DL-aspartic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.
Solubility
Soluble in hot water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
N-Carbamoyl-DL-aspartic acid, is a useful biochemical for proteomics research. It is also used as a carbamate derivative which serves as an intermediate in pyrimidine biosynthesis.
Solubility
Soluble in hot water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
Specifications
Melting Point | ∼175°C (decomposition) |
Beilstein | 1726861 |
Solubility Information | Soluble in hot water. |
Formula Weight | 176.13 |
Percent Purity | 98% |
Quantity | 10 g |
Chemical Name or Material | N-Carbamoyl-DL-aspartic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'