missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Carbobenzyloxy-L-aspartic acid, 98%

Catalog Number p-3757187
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1152-61-0
C12H13NO6
267.23
MFCD00002719
XYXYXSKSTZAEJW-VIFPVBQESA-N
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
2723942
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O
This item is not returnable. View return policy
1152-61-0
C12H13NO6
267.23
MFCD00002719
XYXYXSKSTZAEJW-VIFPVBQESA-N
z-asp-oh, n-carbobenzyloxy-l-aspartic acid, cbz-asp-oh, n-carbobenzoxy-l-aspartic acid, n-cbz-l-aspartic acid, z-l-aspartic acid, carbobenzoxy-l-aspartic acid, n-z-l-aspartic acid, n-benzyloxycarbonylaspartic acid, z-l-asp-oh
2723942
(2S)-2-(phenylmethoxycarbonylamino)butanedioic acid
C1=CC=C(C=C1)COC(=O)NC(CC(=O)O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 117°C to 119°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH(NHCO2CH2C6H5)CO2H
Quantity 25 g
Specific Rotation + 10.5
Solubility Information Solubility in water: .. Other solubilities: soluble in most common organic solvents
Formula Weight 267.23
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material N-Carbobenzyloxy-L-aspartic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'