missing translation for 'onlineSavingsMsg'
Learn More

N-(Chloromethyl)phthalimide, 97%

Product Code. 11449872
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11449872

Brand: Thermo Scientific Alfa Aesar A12018.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17564-64-6 | C9H6ClNO2 | 195.602 g/mol

It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an agrochemical and medicine intermediates and is used in organic synthesis. Microporous membranes prepared from chemically modified polysulfone, which is a derivative of this compound have better streaming potential.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17564-64-6
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00005898
InChI Key JKGLRGGCGUQNEX-UHFFFAOYSA-N
Synonym n-chloromethyl phthalimide, 2-chloromethyl isoindoline-1,3-dione, n-chloromethylphthalimide, chloromethylphthalimide, n-chloromethyltrimellitimide, 1h-isoindole-1,3 2h-dione, 2-chloromethyl, phthalimide, n-chloromethyl, 2-chloromethyl isoindole-1,3-dione, 2-chloromethyl-1h-isoindole-1,3 2h-dione
PubChem CID 87154
IUPAC Name 2-(chloromethyl)isoindole-1,3-dione
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCl

Specifications

Melting Point 131°C to 135°C
Quantity 25 g
Beilstein 140942
Solubility Information Insoluble in water.
Formula Weight 195.61
Percent Purity 97%
Chemical Name or Material N- (Chloromethyl)phthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.