missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Chlorosuccinimide, 97%, Thermo Scientific Chemicals
£20.30 - £208.00
Chemical Identifiers
CAS | 128-09-6 |
---|---|
Molecular Formula | C4H4ClNO2 |
Molecular Weight (g/mol) | 133.53 |
InChI Key | JRNVZBWKYDBUCA-UHFFFAOYSA-N |
Synonym | n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 |
PubChem CID | 31398 |
ChEBI | CHEBI:53203 |
IUPAC Name | 1-chloropyrrolidine-2,5-dione |
SMILES | C1CC(=O)N(C1=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10396640
|
Thermo Scientific Acros
149251000 |
100 g | Plastic bottle |
£20.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697302
|
Thermo Scientific Acros
149255000 |
500 g | Plastic bottle |
£65.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935581
|
Thermo Scientific Acros
149250025 |
2.5 kg | Plastic Bottle |
£208.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
128-09-6 | |
133.53 | |
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
CHEBI:53203 | |
C1CC(=O)N(C1=O)Cl |
C4H4ClNO2 | |
JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
31398 | |
1-chloropyrrolidine-2,5-dione |
Specifications
128-09-6 | |
White | |
4% (2g, 110°C, 2 hrs) max. | |
96% min. (on dry substance) (Iodometry) | |
21,38 | |
15,2167 | |
Solubility in water: 14g/L (20°C). Other solubilities: sparingly soluble in chloroform,ether and ccl4,1g/150mL alcohol- 1g/50mL benzene,soluble in acetone | |
C1CC(=O)N(C1=O)Cl | |
133.53 | |
CHEBI:53203 | |
97% | |
N-Chlorosuccinimide |
144.0°C to 150.0°C | |
>110°C | |
Authentic | |
C4H4ClNO2 | |
01,139; 02,69; 05,127; 06,115; 07,65; 08,103; 09,111; 10,94; 12,121; 13,62; 14,87; 15,86; 16,84 | |
n-chlorosuccinimide, succinchlorimide, chlorosuccinimide, succinochlorimide, succinic n-chloroimide, 2,5-pyrrolidinedione, 1-chloro, 1-chloro-2,5-pyrrolidinedione, succinimide, n-chloro, ncs, caswell no. 807 | |
JRNVZBWKYDBUCA-UHFFFAOYSA-N | |
1-chloropyrrolidine-2,5-dione | |
31398 | |
133.53 | |
Crystalline Powder |
Safety and Handling
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-878-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- N-Chlorosuccinimide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only