missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, 99%

Product Code. p-3345849
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11417776

Brand: Thermo Scientific Alfa Aesar A14732.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 124-18-5 | C10H22 | 142.286 g/mol

n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decane is used as a solvent as well as an internal standard in the GC analysis of oxalic, malonic and succinic acids. It is used in paint manufacture as a hydrocarbon solvent and for wood stains and varnishes as a varnish solvent. It is an active component of automotive fuels viz. gasoline (petrol).

Solubility
Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00008954
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N
Synonym n-decane, nonane, methyl, decyl hydride, unii-nk85062oiy, ccris 653, hsdb 63, dsstox_cid_4913, decane, analytical standard, dsstox_rid_77577, dsstox_gsid_24913
PubChem CID 15600
ChEBI CHEBI:41808
IUPAC Name decane
SMILES CCCCCCCCCC

Specifications

Melting Point °C to 30°C
Density 0.730
Boiling Point 172°C to 174°C
Flash Point 46°C (115°F)
Assay Percent Range 99%
Refractive Index 1.4115
Quantity 100 mL
Beilstein 1696981
Solubility Information Soluble in alcohol and ethers. Miscible with ethanol and ether. Slightly miscible with carbon tetrachloride. Immiscible with water.
Formula Weight 142.29
Physical Form Liquid
Chemical Name or Material n-Decane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Decane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.