missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecane, 99%, pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
250 mL
500 mL
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -12.0°C |
Density | 0.7530g/mL |
Boiling Point | 215.0°C to 217.0°C |
Flash Point | 71°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Molecular Formula | C12H26 |
Linear Formula | CH3(CH2)10CH3 |
Refractive Index | 1.4210 to 1.4230 |
Show More |
Safety and Handling

Product Identifier
- n-Dodecane
- Danger
- Aspiration toxicity Category 1
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction