missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Dodecane, Extra Pure, SLR, Fisher Chemical™
£885.00
Chemical Identifiers
CAS | 112-40-3 |
---|---|
Molecular Formula | C12H26 |
Molecular Weight (g/mol) | 170.34 |
MDL Number | 8969 |
InChI Key | SNRUBQQJIBEYMU-UHFFFAOYSA-N |
Synonym | n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 |
PubChem CID | 8182 |
ChEBI | CHEBI:28817 |
IUPAC Name | dodecane |
SMILES | CCCCCCCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10318393
|
Fisher Chemical
D/5290/17 |
2.5 L | |||||||
Description
Chemical Identifiers
112-40-3 | |
170.34 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
8182 | |
dodecane |
C12H26 | |
8969 | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
CHEBI:28817 | |
CCCCCCCCCCCC |
Specifications
112-40-3 | |
Colorless | |
Amber glass bottle | |
8969 | |
SNRUBQQJIBEYMU-UHFFFAOYSA-N | |
dodecane | |
8182 | |
CHEBI:28817 | |
Liquid |
-12°C | |
215°C | |
C12H26 | |
n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661 | |
CCCCCCCCCCCC | |
170.34 | |
&5.9 | |
170.34g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dodecane
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.