Learn More
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Thermo Scientific Chemicals
For protein solubilization
£66.90 - £169.00
Chemical Identifiers
CAS | 14933-08-5 |
---|---|
Molecular Formula | C17H37NO3S |
Molecular Weight (g/mol) | 335.547 |
MDL Number | MFCD00036909 |
InChI Key | IZWSFJTYBVKZNK-UHFFFAOYSA-N |
Synonym | lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium |
PubChem CID | 84703 |
ChEBI | CHEBI:75303 |
IUPAC Name | 3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate |
SMILES | CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15480537
|
Thermo Scientific Alfa Aesar
J66265.06 |
5 g |
£66.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15490537
|
Thermo Scientific Alfa Aesar
J66265.14 |
25 g |
£169.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For protein solubilization
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14933-08-5 | |
335.547 | |
IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
84703 | |
3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate |
C17H37NO3S | |
MFCD00036909 | |
lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium | |
CHEBI:75303 | |
CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Specifications
14933-08-5 | |
110°C (230°F) | |
MFCD00036909 | |
Hygroscopic | |
Soluble at 1M in water. | |
CCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
335.547 | |
CHEBI:75303 | |
Powder |
White | |
C17H37NO3S | |
4145308 | |
lauryl sultaine, lauryl sulfobetaine, n-dodecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dodecyldimethylammonio propane-1-sulfonate, zwittergent 3-12, 3-dodecyldimethylammonio propanesulfonate, sulfobetaine 12, unii-r6p3kw3e8u, 3-lauryldimethylammonio propanesulfonate, dodecyldimethyl 3-sulphonatopropyl ammonium | |
IZWSFJTYBVKZNK-UHFFFAOYSA-N | |
3-[dodecyl(dimethyl)azaniumyl]propane-1-sulfonate | |
84703 | |
335.55 | |
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 239-002-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only