missing translation for 'onlineSavingsMsg'
Learn More

N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride, 95%, Thermo Scientific Chemicals

£26.00 - £79.10

Chemical Identifiers

CAS 27894-50-4
Molecular Formula C15H23ClN2O4
Molecular Weight (g/mol) 330.81
MDL Number MFCD00034846
InChI Key QPNJISLOYQGQTI-ZOWNYOTGSA-N
Synonym h-lys z-ome.hcl, h-lys z-ome hcl, s-methyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-ome hydrochloride, n epsilon-benzyloxycarbonyl-l-lysine methyl ester hydrochloride, nepsilon-cbz-l-lysine methyl ester hydrochloride, methyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, lys z-ome.hcl, h-lys z-ome?cl, h-lys cbz-och3.hcl
PubChem CID 12935371
IUPAC Name methyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11322247
View Documents
Thermo Scientific Alfa Aesar
L09744.03
1 g
£26.00
1g
Please to purchase this item. Need a web account? Register with us today!
11332247
View Documents
Thermo Scientific Alfa Aesar
L09744.06
5 g
£79.10
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

27894-50-4
330.81
QPNJISLOYQGQTI-ZOWNYOTGSA-N
12935371
Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
C15H23ClN2O4
MFCD00034846
h-lys z-ome.hcl, h-lys z-ome hcl, s-methyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-ome hydrochloride, n epsilon-benzyloxycarbonyl-l-lysine methyl ester hydrochloride, nepsilon-cbz-l-lysine methyl ester hydrochloride, methyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, lys z-ome.hcl, h-lys z-ome?cl, h-lys cbz-och3.hcl
methyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride

Specifications

115°C to 118°C
C15H23ClN2O4
3578476
QPNJISLOYQGQTI-ZOWNYOTGSA-N
methyl (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoate;hydrochloride
12935371
+14.5° (c=2 in Water)
N(epsilon)-Benzyloxycarbonyl-L-lysine methyl ester hydrochloride
27894-50-4
MFCD00034846
h-lys z-ome.hcl, h-lys z-ome hcl, s-methyl 2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, h-lys z-ome hydrochloride, n epsilon-benzyloxycarbonyl-l-lysine methyl ester hydrochloride, nepsilon-cbz-l-lysine methyl ester hydrochloride, methyl 2s-2-amino-6-benzyloxy carbonyl amino hexanoate hydrochloride, lys z-ome.hcl, h-lys z-ome?cl, h-lys cbz-och3.hcl
Cl.COC(=O)[C@@H](N)CCCCNC(=O)OCC1=CC=CC=C1
330.81
330.81
95%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 248-715-9

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only