missing translation for 'onlineSavingsMsg'
Learn More
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals Product Code.: 10637332

N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals

Product Code. 10637332
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10637332

Brand: Thermo Scientific Acros 165350250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16357-59-8
Molecular Formula C14H17NO3
Molecular Weight (g/mol) 247.29
MDL Number MFCD00006703
InChI Key GKQLYSROISKDLL-UHFFFAOYNA-N
Synonym eedq, 2-ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, n-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, ethyl 2-ethoxyquinoline-1 2h-carboxylate, 1-ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 2-ethoxy-1 2h-quinolinecarboxylic acid, ethyl ester, 1 2h-quinolinecarboxylic acid, 2-ethoxy-, ethyl ester, ethyl 2-ethoxy-1,2-dihydroquinoline-1-carboxylate, ethyl 1,2-dihydro-2-ethoxy-1-quinolinecarboxylate, ethyl n-2-ethoxy-1,2-dihydroquinoline carboxylate
PubChem CID 27833
IUPAC Name ethyl 2-ethoxy-2H-quinoline-1-carboxylate
SMILES CCOC1C=CC2=CC=CC=C2N1C(=O)OCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Color White to Beige
Boiling Point 125.0°C to 128.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, V,3, 28
Fieser 02,191; 03,137; 04,223
Merck Index 15, 3566
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 247.29
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonyl-2-ethoxy-1, 2-dihydroquinoline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0436330 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number165350250,165352500,165351000
Lot NumberA0436330 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number165350250,165352500,165351000
Lot NumberA0422934 Certificate TypeCertificate of Analysis Date25/09/2023 Catalog Number165350250,165352500,165351000
Lot NumberA0398719 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number165350250,165352500,165351000
Lot NumberA0399915 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number165350250,165352500,165351000
compliance-icons
Product Identifier
  • N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.