missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)nortropinone, 99%, Thermo Scientific™

£16.17 - £135.68

Chemical Identifiers

CAS 32499-64-2
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00078171
InChI Key ANEJUHJDPGTVIO-UHFFFAOYSA-N
Synonym ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
PubChem CID 401882
IUPAC Name ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CCOC(=O)N1C2CCC1CC(=O)C2
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393277
SDS View Documents
Thermo Scientific Alfa Aesar
L13849.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11303287
SDS View Documents
Thermo Scientific Alfa Aesar
L13849.06
5 g
£40.70
5g
Please to purchase this item. Need a web account? Register with us today!
11313287
SDS View Documents
Thermo Scientific Alfa Aesar
L13849.14
25 g
£135.68
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-(Ethoxycarbonyl)nortropinone, 99%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.