missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethylaniline, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2500 mL
Description
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).
Solubility
Soluble in water (50g/L).
Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
N-Ethylaniline (NEA) may be used as an internal standard in the GC analysis of nicotine extraction from nicotine gum. It may be used in the synthesis of the following, 2-(N-ethylphenylamino)-1,4-benzoquinone, 2-(arylaminomethyl)phenylboronic acid, poly(methyl methacrylate) (PMMA) films containing NEA and poly(N-ethylaniline) (PNEA).
Solubility
Soluble in water (50g/L).
Notes
Air and Light Sensitive, store in dark and away from air. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | -64°C |
Density | 0.961 |
Boiling Point | 204°C to 205°C |
Flash Point | 85°C (185°F) |
Odor | Odorless |
Refractive Index | 1.554 |
Quantity | 500 mL |
UN Number | UN2272 |
Beilstein | 507468 |
Sensitivity | Air and light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'