missing translation for 'onlineSavingsMsg'
Learn More
N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals Product Code.: 11449383

N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals

Product Code. 11449383
100 mL
Click to view available options
Quantity:
100 mL
500 mL

Product Code. 11449383

Brand: Thermo Scientific Alfa Aesar A11801.AE

Please to purchase this item. Need a web account? Register with us today!

N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethyldiisopropylamine is used to make or formulate industrial products. It is also used in organic chemistry as a base. It acts as a selective reagent in the alkylation of secondary amines to tertiary amines by alkyl halides. It reacts with monoactivated Michael acceptors to afford symmetrical sulfones. Further, it is used to prepare heterocyclic compound, sscorpionine by a reaction with disulfur dichloride using 1,4-diazabicyclo[2.2.2]octane (DABCO) as a catalyst.

Solubility
Miscible with water and ethanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides, carbon dioxide, copper, brass, rubber, oxidizing agents, nitrates, peroxides, aldehydes and metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7087-68-5
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008868
InChI Key JGFZNNIVVJXRND-UHFFFAOYSA-N
Synonym n,n-diisopropylethylamine, ethyldiisopropylamine, n-ethyldiisopropylamine, diisopropylethylamine, diea, hunig's base, n-ethyl-n-isopropylpropan-2-amine, dipea, 2-propanamine, n-ethyl-n-1-methylethyl, 1,1'-dimethyltriethylamine
PubChem CID 81531
IUPAC Name N-ethyl-N-propan-2-ylpropan-2-amine
SMILES CCN(C(C)C)C(C)C

Specifications

Melting Point −46°C
Density 0.747
Boiling Point 126°C to 127°C
Flash Point 8°C (46°F)
Assay Percent Range 99%
Refractive Index 1.4135
Quantity 100 mL
Beilstein 605301
Solubility Information Miscible with water and ethanol.
Formula Weight 129.25
Physical Form Liquid
Chemical Name or Material N-Ethyldiisopropylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Ethyldiisopropylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : N-Ethyldiisopropylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Ethyldiisopropylamine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.