missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylethylenediamine, 98%, Thermo Scientific Chemicals

£18.20 - £56.80

Chemical Identifiers

CAS 110-72-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008166
InChI Key SCZVXVGZMZRGRU-UHFFFAOYSA-N
Synonym n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
PubChem CID 66071
IUPAC Name N'-ethylethane-1,2-diamine
SMILES CCNCCN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435607
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23577.06
5 g
£18.20
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11445607
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23577.14
25 g
£56.80
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylethylenediamine is a chelating agent used in mutagenesis research studies. It undergoes condensation with 2-hydroxy-3-methoxybenzaldehyde. It also forms organometallic ruthenium(II) complexes that inhibit the growth in human ovarian cancer cell lines.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
Specifications

Chemical Identifiers

110-72-5
88.154
SCZVXVGZMZRGRU-UHFFFAOYSA-N
66071
CCNCCN
C4H12N2
MFCD00008166
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
N'-ethylethane-1,2-diamine

Specifications

110-72-5
128°C to 130°C
C4H12N2
MFCD00008166
1098272
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
CCNCCN
88.154
88.15
N-Ethylethylenediamine
0.837
34°C (93°F)
1.44
UN2733
Air Sensitive
SCZVXVGZMZRGRU-UHFFFAOYSA-N
N'-ethylethane-1,2-diamine
66071
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H225-H314

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II

missing translation for 'einecsNumber' : 203-795-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only