Learn More
N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals
£53.80 - £1186.00
Chemical Identifiers
CAS | 128-53-0 |
---|---|
Molecular Formula | C6H7NO2 |
Molecular Weight (g/mol) | 125.13 |
InChI Key | HDFGOPSGAURCEO-UHFFFAOYSA-N |
Synonym | n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide |
PubChem CID | 4362 |
ChEBI | CHEBI:44485 |
IUPAC Name | 1-ethylpyrrole-2,5-dione |
SMILES | CCN1C(=O)C=CC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10266200
|
Thermo Scientific Acros
156100050 |
5 g |
£53.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10021790
|
Thermo Scientific Acros
156100100 |
10 g |
£89.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10112970
|
Thermo Scientific Acros
156100500 |
50 g |
£320.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11945581
|
Thermo Scientific Acros
156102500 |
250 g |
£1186.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
128-53-0 | |
125.13 | |
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide | |
CHEBI:44485 | |
CCN1C(=O)C=CC1=O |
C6H7NO2 | |
HDFGOPSGAURCEO-UHFFFAOYSA-N | |
4362 | |
1-ethylpyrrole-2,5-dione |
Specifications
128-53-0 | |
White | |
73°C | |
99% min. (GC) | |
C6H7NO2 | |
15,3877 | |
Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C) | |
CCN1C(=O)C=CC1=O | |
125.13 | |
CHEBI:44485 | |
99+% | |
N-Ethylmaleimide |
41.0°C to 45.0°C | |
210.0°C | |
Authentic | |
Glass bottle | |
21,IV,4629 | |
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide | |
HDFGOPSGAURCEO-UHFFFAOYSA-N | |
1-ethylpyrrole-2,5-dione | |
4362 | |
125.13 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-892-4
RUO – Research Use Only