missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals

£58.19 - £186.93

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amineShow MoreShow Less
PubChem CID 12219
IUPAC Name N-methylethanamine
SMILES CCNC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376916
SDS View Documents
Thermo Scientific Alfa Aesar
L04372.06
5 g
£58.19
5g
Please to purchase this item. Need a web account? Register with us today!
11386916
SDS View Documents
Thermo Scientific Alfa Aesar
L04372.14
25 g
£186.93
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

missing translation for 'einecsNumber' : 210-862-1

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.