missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylphthalimide, 98%, Thermo Scientific Chemicals

Product Code. 11428636
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11428636

Brand: Thermo Scientific Alfa Aesar A16014.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5022-29-7
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00014583
InChI Key JZDSOQSUCWVBMV-UHFFFAOYSA-N
Synonym n-ethylphthalimide, 2-ethylisoindoline-1,3-dione, phthalimide, n-ethyl, 1h-isoindole-1,3 2h-dione, 2-ethyl, 2-ethyl-1h-isoindole-1,3 2h-dione, 1h-isoindole-1,3 2h-dione, n-ethyl, 2-ethylbenzo c azoline-1,3-dione, acmc-1az90, 1h-isoindole-1, n-ethyl
PubChem CID 21120
IUPAC Name 2-ethylisoindole-1,3-dione
SMILES CCN1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 74°C to 78°C
Quantity 5 g
Beilstein 132094
Formula Weight 175.19
Percent Purity 98%
Chemical Name or Material N-Ethylphthalimide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.