missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-3-(2-naphthyl)-L-alanine, 95%, Thermo Scientific Chemicals
£31.50 - £218.00
Chemical Identifiers
CAS | 112883-43-9 |
---|---|
Molecular Formula | C28H22NO4 |
Molecular Weight (g/mol) | 436.49 |
MDL Number | MFCD00144886 |
InChI Key | JYUTZJVERLGMQZ-SANMLTNESA-M |
Synonym | fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347 |
PubChem CID | 2734452 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid |
SMILES | [O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438737
|
Thermo Scientific Alfa Aesar
H52179.MD |
250 mg |
£31.50
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15418737
|
Thermo Scientific Alfa Aesar
H52179.03 |
1 g |
£79.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15428737
|
Thermo Scientific Alfa Aesar
H52179.06 |
5 g |
£218.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
112883-43-9 | |
436.49 | |
JYUTZJVERLGMQZ-SANMLTNESA-M | |
2734452 | |
[O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C28H22NO4 | |
MFCD00144886 | |
fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347 | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid |
Specifications
112883-43-9 | |
MFCD00144886 | |
fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347 | |
JYUTZJVERLGMQZ-SANMLTNESA-M | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid | |
2734452 | |
95% |
C28H22NO4 | |
7668973 | |
Slightly soluble in water. | |
[O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
436.49 | |
437.49 | |
N-Fmoc-3-(2-naphthyl)-L-alanine |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only