missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(3-pyridyl)-L-alanine, 95%, Thermo Scientific Chemicals

Product Code. 15438557
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175453-07-3
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00144887
InChI Key JQLPMTXRCLXOJO-NRFANRHFSA-N
Synonym fmoc-3-3-pyridyl-l-alanine, fmoc-l-3-pyridylalanine, fmoc-3-pal-oh, fmoc-beta-3-pyridyl-ala-oh, fmoc-l-3-3-pyridyl-alanine, fmoc-3-pyridyl-ala-oh, fmoc-3'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid, n-fmoc-3-3-pyridyl-l-alanine, fmoc-d-3-3-pyridyl-alanine
PubChem CID 6957974
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O

Specifications

Quantity 250 mg
Beilstein 9154190
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(3-pyridyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'