missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-iodo-L-phenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15408457
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82565-68-2
Molecular Formula C24H20INO4
Molecular Weight (g/mol) 513.33
MDL Number MFCD00672340
InChI Key LXOXXTQKKRJNNB-QFIPXVFZSA-N
Synonym fmoc-4-iodo-l-phenylalanine, fmoc-phe 4-i-oh, fmoc-l-4-iodophenylalanine, fmoc-l-phe 4-i-oh, fmoc-p-iodo-phe-oh, fmoc-4'-iodo-l-phe, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-iodophenyl propanoic acid, fmoc-l-phe 4-i, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-iodophenyl propanoic acid, l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-iodo
PubChem CID 2761479
SMILES OC(=O)[C@H](CC1=CC=C(I)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 213°C to 217°C
Quantity 250 mg
Sensitivity Light sensitive
Optical Rotation −10° (c=1 in DMF)
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-iodophenyl)propanoic acid
Formula Weight 513.33
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-iodo-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'