missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-nitro-L-phenylalanine, 98%, Thermo Scientific Chemicals

£40.50 - £143.00

Chemical Identifiers

CAS 95753-55-2
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD00057810
InChI Key RZRRJPNDKJOLHI-QFIPXVFZSA-N
Synonym fmoc-4-nitro-l-phenylalanine, fmoc-phe 4-no2-oh, fmoc-l-4-no2-phe-oh, fmoc-p-nitro-phe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-nitrophenyl propanoic acid, n-fmoc-4-nitro-l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-nitro-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-nitrophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-3-4-nitrophenyl propanoic acid, fmoc-phe no2-oh
PubChem CID 7016054
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15498417
View Documents
Thermo Scientific Alfa Aesar
H51962.03
1 g
£40.50
1g
Please to purchase this item. Need a web account? Register with us today!
15408427
View Documents
Thermo Scientific Alfa Aesar
H51962.06
5 g
£143.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95753-55-2
432.43
RZRRJPNDKJOLHI-QFIPXVFZSA-N
7016054
OC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C24H20N2O6
MFCD00057810
fmoc-4-nitro-l-phenylalanine, fmoc-phe 4-no2-oh, fmoc-l-4-no2-phe-oh, fmoc-p-nitro-phe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-nitrophenyl propanoic acid, n-fmoc-4-nitro-l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-nitro-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-nitrophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-3-4-nitrophenyl propanoic acid, fmoc-phe no2-oh
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid

Specifications

95753-55-2
C24H20N2O6
5178656
RZRRJPNDKJOLHI-QFIPXVFZSA-N
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid
7016054
98%
213°C to 223°C
MFCD00057810
fmoc-4-nitro-l-phenylalanine, fmoc-phe 4-no2-oh, fmoc-l-4-no2-phe-oh, fmoc-p-nitro-phe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-nitrophenyl propanoic acid, n-fmoc-4-nitro-l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-nitro-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-nitrophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-3-4-nitrophenyl propanoic acid, fmoc-phe no2-oh
OC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
432.43
432.43
N-Fmoc-4-nitro-L-phenylalanine
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only