missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-D-glutamic acid 5-tert-butyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 83°C to 89°C |
Solubility Information | Slightly soluble in water. |
Formula Weight | 425.48 |
Percent Purity | 98% |
Quantity | 25 g |
Physical Form | Solid |
Chemical Name or Material | N-Fmoc-D-glutamic acid 5-tert-butyl ester |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'