missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-pipecolic acid, ≥96%, Thermo Scientific™

Product Code. 15463928
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101555-63-9
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD00235899
InChI Key CKLAZLINARHOTG-LJQANCHMSA-N
Synonym fmoc-d-pipecolic acid, fmoc-d-pip-oh, fmoc-d-hopro-oh, d-1-fmoc-pipecolinic acid, fmoc-d-piperidine-2-carboxylic acid, fmoc-d-homopro-oh, d-n-fmoc-pipecolic acid, r-n-fmoc-piperidine-2-carboxylic acid, r-1-9h-fluoren-9-yl methoxy carbonyl piperidine-2-carboxylic acid, fmoc-d-homoproline
PubChem CID 6958378
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)piperidine-2-carboxylic acid
SMILES C1CCN(C(C1)C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Melting Point 154°C to 159°C
Color Brown to White
Quantity 250 mg
Formula Weight 351.4
Percent Purity ≥96%
Physical Form Solid
Chemical Name or Material N-Fmoc-D-pipecolic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'