missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-serine, 95%, Thermo Scientific Chemicals

Product Code. 15479008
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116861-26-8
Molecular Formula C18H17NO5
Molecular Weight (g/mol) 327.336
MDL Number MFCD00077068
InChI Key JZTKZVJMSCONAK-MRXNPFEDSA-N
Synonym fmoc-d-ser-oh, fmoc-d-serine, n-9h-fluoren-9-ylmethoxy carbonyl-d-serine, n-fmoc-d-serine, d-serine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-serine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxypropanoic acid, ambotzfaa1508, pubchem10038, n-fmoc-d-ala oh
PubChem CID 688637
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxypropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CO)C(=O)O

Specifications

Melting Point 108°C to 112°C
Quantity 5 g
Beilstein 4715790
Optical Rotation +12° (c=1 in DMF)
Formula Weight 327.33
Percent Purity 95%
Chemical Name or Material N-Fmoc-D-serine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'