missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-glycine, 98%, Thermo Scientific Chemicals

Product Code. 11430147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29022-11-5
Molecular Formula C17H15NO4
Molecular Weight (g/mol) 297.31
MDL Number MFCD00037140
InChI Key NDKDFTQNXLHCGO-UHFFFAOYSA-N
Synonym fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
PubChem CID 93124
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 172°C to 175°C
Quantity 5 g
Beilstein 2163967
Formula Weight 297.32
Percent Purity 98%
Chemical Name or Material N-Fmoc-glycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'