missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-alanine monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 11484567
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207291-76-7
Molecular Formula C18H19NO5
Molecular Weight (g/mol) 329.35
MDL Number MFCD00037139
InChI Key GAPWKFLOMOFHGO-UHFFFAOYNA-N
Synonym n-fmoc-l-alanine hydrate, n-fmoc-l-alanine monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-alanine hydrate, n-fmoc-l-alanine, 2s-2-fluoren-9-ylmethoxy carbonylamino propanoic acid, hydrate, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid hydrate, fmoc-ala-oh h2o, fmoc-ala-oh.h2o, fmoc-l-alanine hydrate, n-9-fluorenylmethoxycarbonyl-l-alanine monohydrate
PubChem CID 18528118
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid;hydrate
SMILES O.CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point ∼130°C
Quantity 25 g
Beilstein 2225975
Solubility Information Insoluble in water.
Optical Rotation −19° (c=1 in DMF)
Formula Weight 329.36 (311.34 Anhydrous)
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-alanine monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'