missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-threonine monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 11474597
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73731-37-0
Molecular Formula C19H21NO6
Molecular Weight (g/mol) 359.38
MDL Number MFCD00792901
InChI Key CGNQPFAECJFQNV-NRNQBQMASA-N
Synonym fmoc-thr-oh, fmoc-l-threonine, fmoc-l-thr-oh, 2-9h-fluoren-9-ylmethoxycarbonylamino-3-hydroxy-butanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-hydroxybutanoic acid, n-9-fluorenylmethoxycarbonyl-l-threonine, 2s,3r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-hydroxybutanoic acid, fmoc-thr-oh monohydrate, n-9h-fluoren-9-ylmethoxy carbonyl-l-threonine monohydrate
PubChem CID 6992530
IUPAC Name (2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-hydroxybutanoic acid
SMILES O.C[C@@H](O)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 94°C to 96°C
Quantity 1 g
Optical Rotation −16° (c=1 in DMF)
Formula Weight 359.38 (341.36 Anhydrous)
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-threonine monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'