Learn More
n-Heptane, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
£34.77 - £67.84
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10396591
|
Thermo Scientific Acros
326911000 |
100 mL |
£34.77
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10580994
|
Thermo Scientific Acros
326910010 |
1 L |
£67.84
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-91.0°C | |
99.0 | |
100.0 | |
98.0°C | |
Authentic | |
1.3850 to 1.3890 | |
0.0005 % max. | |
C7H16 | |
MFCD00009544 | |
01, 154 | |
15, 4694 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
≥99% | |
n-Heptane, Water <30ppm |
142-82-5 | |
Colorless | |
0.6830g/mL | |
−4°C | |
Liquid | |
99+% | |
0.683 | |
CH3(CH2)5CH3 | |
0.4 mPa.s (20°C) | |
0.005% Max. | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2 | |
Extra Dry |
Safety and Handling
GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Danger
missing translation for 'einecsNumber' : 205-563-8
missing translation for 'rtecsNumber' : MI7700000
missing translation for 'tsca' : TSCA
- n-Heptane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.