missing translation for 'onlineSavingsMsg'
Learn More
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™ Product Code.: 10598800

n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™

Product Code. 10598800
2.5 L, Amber Glass Bottle
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10598800

Brand: Fisher Chemical H/0106/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 197-300nm range, to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Filtered through a sub-micron filter to remove particulates
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-82-5
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00009544
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
PubChem CID 8900
ChEBI CHEBI:43098
IUPAC Name heptane
SMILES CCCCCCC

Specifications

Melting Point -91°C
Color Colorless
Boiling Point 98°C
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 48 mbar at 20°C
Viscosity 0.4 mPaS at 20°C
Formula Weight 100.2g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2527693 Certificate TypeCertificate of Analysis Date20/02/2025 Catalog NumberH/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17
Lot Number2527346 Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberH/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17
Lot Number2527159 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberH/0106/30RSS,H/0106/PB17,H/0106/25RSS,H/0106/17X,H/0106/15,H/0106/17Z,H/0106/10RSS,H/0106/15X,H/0106/17
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Heptane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : n-Heptane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.