missing translation for 'onlineSavingsMsg'
Learn More

n-Heptyl 4-hydroxybenzoate, 98%, Thermo Scientific Chemicals

Catalog Number 11490044
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1085-12-7
C14H20O3
236.31
MFCD00016481
ZTJORNVITHUQJA-UHFFFAOYSA-N
heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester
14138
CHEBI:34788
heptyl 4-hydroxybenzoate
CCCCCCCOC(=O)C1=CC=C(O)C=C1
This item is not returnable. View return policy
1085-12-7
C14H20O3
236.31
MFCD00016481
ZTJORNVITHUQJA-UHFFFAOYSA-N
heptylparaben, heptyl paraben, heptyl p-hydroxybenzoate, n-heptyl 4-hydroxybenzoate, nipaheptyl, staypro ws 7, n-heptyl p-hydroxybenzoate, benzoic acid, 4-hydroxy-, heptyl ester, p-hydroxybenzoic acid heptyl ester, p-oxybenzoesaureheptylester
14138
CHEBI:34788
heptyl 4-hydroxybenzoate
CCCCCCCOC(=O)C1=CC=C(O)C=C1

n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.

Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 46°C to 49°C
Quantity 25 g
Solubility Information Soluble in alcohol,phenoxyethanol,propylene glycol. Insoluble in water.
Formula Weight 236.31
Percent Purity 98%
Chemical Name or Material n-Heptyl 4-hydroxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'