Learn More
n-Heptylamine, 99+%, Thermo Scientific Chemicals
£23.82 - £212.06
Chemical Identifiers
CAS | 111-68-2 |
---|---|
Molecular Formula | C7H17N |
Molecular Weight (g/mol) | 115.22 |
MDL Number | MFCD00008244 |
InChI Key | WJYIASZWHGOTOU-UHFFFAOYSA-N |
Synonym | heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 |
PubChem CID | 8127 |
IUPAC Name | heptan-1-amine |
SMILES | CCCCCCCN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071690
|
Thermo Scientific Acros
155460250 |
25 mL | Glass bottle |
£23.82
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10081690
|
Thermo Scientific Acros
155461000 |
100 mL | Glass bottle |
£60.20
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15179193
|
Thermo Scientific Acros
155465000 |
500 mL | Glass bottle |
£212.06
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-68-2 | |
115.22 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
8127 | |
CCCCCCCN |
C7H17N | |
MFCD00008244 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
heptan-1-amine |
Specifications
111-68-2 | |
100.0 | |
Colorless to Yellow | |
153.0°C to 156.0°C | |
Authentic | |
C7H17N | |
1.4233 to 1.4253 | |
04,193 | |
heptylamine, 1-aminoheptane, 1-heptanamine, n-heptylamine, 1-heptylamine, heptanamine, heptyl-amine, heptyl amine, dsstox_cid_681 | |
WJYIASZWHGOTOU-UHFFFAOYSA-N | |
heptan-1-amine | |
8127 | |
99+% | |
n-Heptylamine, 99+% |
99.0 | |
-23.0°C | |
0.7740g/mL | |
44°C | |
99+% | |
CH3(CH2)6NH2 | |
MFCD00008244 | |
0.774 | |
Solubility in water: 100g/L water (20°C). Other solubilities: soluble in alcohol and ether | |
CCCCCCCN | |
115.22 | |
115.22 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-895-8
missing translation for 'rtecsNumber' : MK0600000
missing translation for 'tsca' : TSCA
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 C
- Skin sensitiser Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only