missing translation for 'onlineSavingsMsg'
Learn More

n-Hexacosane, 99%, Thermo Scientific Chemicals

Product Code. 11318978
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-01-3
Molecular Formula C26H54
Molecular Weight (g/mol) 366.72
MDL Number MFCD00009354
InChI Key HMSWAIKSFDFLKN-UHFFFAOYSA-N
Synonym n-hexacosane, pentacosane, methyl, ch3-ch2 24-ch3, hexacosane, analytical standard, hexacosane, acmc-1b86y
PubChem CID 12407
ChEBI CHEBI:32940
IUPAC Name hexacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 56°C to 58°C
Color White
Odor Weak
Quantity 25 g
Beilstein 1705609
Solubility Information Slightly soluble in water.
Formula Weight 366.72
Percent Purity 99%
Physical Form Solid
Chemical Name or Material n-Hexacosane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'