missing translation for 'onlineSavingsMsg'
Learn More
n-Hexadecane, >99%, MP Biomedicals™ Product Code.: 11411142
compliance-icons

n-Hexadecane, >99%, MP Biomedicals™

Product Code. 11411142 Shop All MP Biomedicals Products
500 mL
Click to view available options
Quantity:
500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11411142

Brand: MP Biomedicals 0219521890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

n-Hexadecane ≥99%, Liquid

  • Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP. Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a reference fuel with the cetane number 100 and is employed in determining the cetane number of diesel fuels. The mechanism of the oxidation of hexadecane in the gas-phase has been described. The density, speed of sound, adiabatic compressibility and compressed liquid density of hexadecane have been determined. The performance of platinum/zeolite catalysts for the hydroisomerization of n-hexadecane has been assessed. The thermal degradation of n-hexadecane forms n-alkanes and 1-alkenes as key products.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-76-3
Molecular Formula C16H34
Molecular Weight (g/mol) 226.448
MDL Number MFCD00008998
InChI Key DCAYPVUWAIABOU-UHFFFAOYSA-N
PubChem CID 11006
ChEBI CHEBI:45296
IUPAC Name hexadecane
SMILES CCCCCCCCCCCCCCCC

Specifications

Color Colorless to White
Density 0.77g/mL
Assay Percent Range >99%
Quantity 500 mL
Synonym n-hexadecane, cetane, n-cetane, hexadecan, hexadekan, cetan, zetan, unii-f8z00shp6q, ccris 5833, pentadecane, methyl
Formula Weight 226.4
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material n-Hexadecane
compliance-icons
Product Identifier
  • N-Hexadecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : n-Hexadecane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexadecane, >99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.