missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane 95%, ChromAR™, Macron Fine Chemicals™
Click to view available options
2.5 L
Specifications
Specifications
CAS | 110-54-3 |
Molecular Formula | C6H14 |
MDL Number | MFCD02179311 |
UN Number | 1208 |
InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
SMILES | CCCCCC |
IUPAC Name | hexane |
Molecular Weight (g/mol) | 86.18 |
Quantity | 2.5 L |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'