missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Hexane, 95%, Optima™ for HPLC and GC/MS, Fisher Chemical™
74.28 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass
Safe-Cote™ Amber Glass
Description
- Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
- Tested for GC/MS suitability
- Also meets ACS specifications
- Intended for research use only
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -95°C |
Absorbance | 0.005 AU max. at 254nm, 0.005 AU max. at 280nm, 0.07 AU max. at 220nm, 0.25 AU max. at 210nm, 1.00 AU max. at 195nm |
Color | Colorless |
Density | 0.659g/cm³ |
Boiling Point | 69°C |
Quantity | 1 L |
Identification | Pass Test |
Packaging | Amber Soda Lime Glass |
Physical Form | Liquid |
Refractive Index | 1.3710 to 1.3750 (at 25°C) |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS08_Dangerous_Health_Hazard.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Product Identifier
- n-Hexane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P331-Do NOT induce vomiting.
- MIXTURE LIST-Contains : hexane, 2-methylpentane, 3-methylpentane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'