missing translation for 'onlineSavingsMsg'
Learn More
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals Product Code.: 15743146
compliance-icons compliance-icons compliance-icons compliance-icons

n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals

Product Code. 15743146
2.5 L
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 15743146

Brand: Thermo Scientific Acros 445720025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym gettysolve-b, n-hexane, hexyl hydride, esani, heksan, skellysolve b, hexan, hexanes, dipropyl, hexanen
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95.0°C
Density 0.6590g/mL
Boiling Point 69.0°C
Flash Point −22°C
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Linear Formula CH3(CH2)4CH3
Refractive Index 1.3748 to 1.3810
Quantity 2.5 L
Merck Index 15,4732
Specific Gravity 0.659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether and chloroform
Viscosity 0.31 mPa.s (20°C)
Formula Weight 86.18
Percent Purity 97+%
Grade HPLC
Chemical Name or Material n-Hexane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2401502 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number445720010,445720025
Lot Number2412288 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number445720010,445720025
Lot Number2496512 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number445720010,445720025
Lot Number1904281 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number445720010,445720025
Lot Number1992271 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number445720010,445720025
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • n-Hexane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H315-Causes skin irritation.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
n-Hexane, 97+%, for HPLC, ACS reagent, meets the requirm. of Reag. Ph.Eur., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.